2-{[(4-Methoxyphenyl)amino]methylidene}cyclohexane-1,3-dione, tech

Lieferant: Apollo Scientific

OR32098-1G
APOSOR32098-1GEA 1570 EUR
APOSOR32098-1G
2-{[(4-Methoxyphenyl)amino]methylidene}cyclohexane-1,3-dione, tech
2-{[(4-Methoxyphenyl)amino]methylidene}cyclohexane-1,3-dione
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01569055
CAS-Nummer: 62370-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR