1-(4-chloro-2-nitrophenyl)piperazine
Lieferant: Apollo Scientific
|
Achtung
|
OR3210-250MG
APOSOR3210-250MGEA
0
EUR
APOSOR3210-250MG
1-(4-chloro-2-nitrophenyl)piperazine
1-(4-chloro-2-nitrophenyl)piperazine
Formel:
C₁₀H₁₂ClN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00640777 CAS-Nummer: 405910-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...