3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-propyl-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

OR32100-1G
APOSOR32100-1GEA 1570 EUR
APOSOR32100-1G
3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-propyl-1,2-dihydropyridin-2-one, tech
3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-propyl-1,2-dihydropyridin-2-one
Formel: C₁₅H₁₆ClNO₂
Molecular Weight: 277,746 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568725
CAS-Nummer: 477846-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR