2-Chlor-6-methoxybenzoesäure

Lieferant: Apollo Scientific

OR321002-5G
APOSOR321002-5GEA 115 EUR
APOSOR321002-5G
2-Chlor-6-methoxybenzoesäure
2-Chloro-6-methoxybenzoic acid
Formel: C₈H₇ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00127702
CAS-Nummer: 3260-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR