3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-(2-methoxyethyl)-1,2-dihydropyridin-2-one, tech
Lieferant: Apollo Scientific
OR32101-1G
APOSOR32101-1GEA
1570
EUR
APOSOR32101-1G
3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-(2-methoxyethyl)-1,2-dihydropyridin-2-one, tech
3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-(2-methoxyethyl)-1,2-dihydropyridin-2-one
Formel:
C₁₅H₁₆ClNO₃ Molecular Weight: 293,745 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01871393 CAS-Nummer: 477846-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...