Methyl-2-amino-4-methylbenzoate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-amino-4-methylbenzoate, 2-Amino-4-methylbenzoic acid methyl ester
OR321010-1G
APOSOR321010-1GEA
62.5
EUR
APOSOR321010-1G
Methyl-2-amino-4-methylbenzoate 98%
Methyl-2-amino-4-methylbenzoate
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09998260 CAS-Nummer: 18595-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...