(2E)-3-(Dimethylamino)-1-(7-methoxy-1-benzofuran-2-yl)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32109-1G
APOSOR32109-1GEA 1570 EUR
APOSOR32109-1G
(2E)-3-(Dimethylamino)-1-(7-methoxy-1-benzofuran-2-yl)prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-(7-methoxy-1-benzofuran-2-yl)prop-2-en-1-one
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082055
CAS-Nummer: 477848-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR