Methyl 4-cyclohexanecarbonyl-1H-pyrrole-2-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl-4-(cyclohexylcarbonyl)-1H-pyrrole-2-carboxylate
OR32114-1G
APOSOR32114-1GEA
671
EUR
APOSOR32114-1G
Methyl 4-cyclohexanecarbonyl-1H-pyrrole-2-carboxylate 95%
Methyl 4-cyclohexanecarbonyl-1H-pyrrole-2-carboxylate
Formel:
C₁₃H₁₇NO₃ Molecular Weight: 235,279 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02571975 CAS-Nummer: 477848-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...