Methyl (2E)-3-(3,4-dimethoxyphenyl)prop-2-enoate, tech

Lieferant: Apollo Scientific

OR32115-1G
APOSOR32115-1GEA 1570 EUR
APOSOR32115-1G
Methyl (2E)-3-(3,4-dimethoxyphenyl)prop-2-enoate, tech
Methyl (2E)-3-(3,4-dimethoxyphenyl)prop-2-enoate
Formel: C₁₂H₁₄O₄
Molecular Weight: 222,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00458425
CAS-Nummer: 30461-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR