Methyl (2E)-3-(3-bromophenyl)prop-2-enoate 95%
Lieferant: Apollo Scientific
Synonyme:
(E)-Methyl 3-(3-bromophenyl)acrylate
OR32116-5G
APOSOR32116-5GEA
214
EUR
APOSOR32116-5G
Methyl (2E)-3-(3-bromophenyl)prop-2-enoate 95%
Methyl (2E)-3-(3-bromophenyl)prop-2-enoate
Formel:
C₁₀H₉BrO₂ Molecular Weight: 241,081 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00296965 CAS-Nummer: 79432-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...