4-Methoxy-3-(2-propynyloxy)benzenecarbaldehyde 95%

Lieferant: Apollo Scientific

OR32119-1G OR32119-5G
APOSOR32119-1GEA 0 EUR
APOSOR32119-1G APOSOR32119-5G
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehyde 95%
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehyde
Formel: C₁₁H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03001224
CAS-Nummer: 145654-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR