2-(3-Chlorophenyl)-5-hydroxy-2,3-dihydropyridazin-3-one, tech

Lieferant: Apollo Scientific

OR32120-1G
APOSOR32120-1GEA 1570 EUR
APOSOR32120-1G
2-(3-Chlorophenyl)-5-hydroxy-2,3-dihydropyridazin-3-one, tech
2-(3-Chlorophenyl)-5-hydroxy-2,3-dihydropyridazin-3-one
Formel: C₁₀H₇ClN₂O₂
Molecular Weight: 222,628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03011979
CAS-Nummer: 383147-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR