tert-Butyl (R)-2-benzylpiperidine-1-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321202-250MG
APOSOR321202-250MGEA
250
EUR
APOSOR321202-250MG
tert-Butyl (R)-2-benzylpiperidine-1-carboxylate 97%
tert-Butyl (R)-2-benzylpiperidine-1-carboxylate
Formel:
C₁₇H₂₅NO₂ Molecular Weight: 275,386 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28007734 CAS-Nummer: 1242146-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...