tert-Butyl (S)-2-benzylpiperidine-1-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321203-100MG
APOSOR321203-100MGEA
250
EUR
APOSOR321203-100MG
tert-Butyl (S)-2-benzylpiperidine-1-carboxylate 97%
tert-Butyl (S)-2-benzylpiperidine-1-carboxylate
Formel:
C₁₇H₂₅NO₂ Molecular Weight: 275,386 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28355902 CAS-Nummer: 1853230-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...