tert-Butyl (R)-2-ethylpiperidine-1-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321205-100MG
APOSOR321205-100MGEA
82.8
EUR
APOSOR321205-100MG
tert-Butyl (R)-2-ethylpiperidine-1-carboxylate 97%
tert-Butyl (R)-2-ethylpiperidine-1-carboxylate
Formel:
C₁₂H₂₃NO₂ Molecular Weight: 213,317 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18832116 CAS-Nummer: 664364-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...