(R)-2-Propylpiperidine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321207-100MG
APOSOR321207-100MGEA 134 EUR
APOSOR321207-100MG
(R)-2-Propylpiperidine hydrochloride 97%
(R)-2-Propylpiperidine hydrochloride
Formel: C₈H₁₇N·ClH
Molecular Weight: 163,688 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03643712
CAS-Nummer: 88057-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR