Ethyl (R)-2-(piperidin-2-yl)acetate hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321209-100MG
APOSOR321209-100MGEA 168 EUR
APOSOR321209-100MG
Ethyl (R)-2-(piperidin-2-yl)acetate hydrochloride 97%
Ethyl (R)-2-(piperidin-2-yl)acetate hydrochloride
Formel: C₉H₁₇NO₂·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD13184088
CAS-Nummer: 1000291-88-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR