Ethyl 2-(naphthalen-1-yloxy)acetate, tech
Lieferant: Apollo Scientific
OR32121-1G
APOSOR32121-1GEA
1570
EUR
APOSOR32121-1G
Ethyl 2-(naphthalen-1-yloxy)acetate, tech
Ethyl 2-(naphthalen-1-yloxy)acetate
Formel:
C₁₄H₁₄O₃ Molecular Weight: 230,259 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01162455 CAS-Nummer: 41643-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...