2-(1,2,3,6-Tetrahydropyridin-4-yl)pyrazine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321213-100MG
APOSOR321213-100MGEA 137 EUR
APOSOR321213-100MG
2-(1,2,3,6-Tetrahydropyridin-4-yl)pyrazine dihydrochloride 97%
2-(1,2,3,6-Tetrahydropyridin-4-yl)pyrazine dihydrochloride
Formel: C₉H₁₁N₃·2ClH
Molecular Weight: 234,126 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075570
CAS-Nummer: 2140866-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR