N-(4-(1,2,3,6-Tetrahydropyridin-4-yl)phenyl)acetamide hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321214-100MG
APOSOR321214-100MGEA 137 EUR
APOSOR321214-100MG
N-(4-(1,2,3,6-Tetrahydropyridin-4-yl)phenyl)acetamide hydrochloride 97%
N-(4-(1,2,3,6-Tetrahydropyridin-4-yl)phenyl)acetamide hydrochloride
Formel: C₁₃H₁₆N₂O·ClH
Molecular Weight: 252,74 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075571
CAS-Nummer: 1853217-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR