5-(1,2,3,6-Tetrahydropyridin-4-yl)thiazole hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321216-100MG
APOSOR321216-100MGEA 168 EUR
APOSOR321216-100MG
5-(1,2,3,6-Tetrahydropyridin-4-yl)thiazole hydrochloride 97%
5-(1,2,3,6-Tetrahydropyridin-4-yl)thiazole hydrochloride
Formel: C₈H₁₀N₂S·ClH
Molecular Weight: 202,704 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075569
CAS-Nummer: 2244084-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR