4-(3,4,5-Trichlorophenyl)piperidine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321219-100MG
APOSOR321219-100MGEA 333 EUR
APOSOR321219-100MG
4-(3,4,5-Trichlorophenyl)piperidine hydrochloride 97%
4-(3,4,5-Trichlorophenyl)piperidine hydrochloride
Formel: C₁₁H₁₂Cl₃N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD29075578
CAS-Nummer: 1853217-62-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR