N-(2-Methylnaphthalen-1-yl)methanesulphonamide, tech

Lieferant: Apollo Scientific

OR32122-1G
APOSOR32122-1GEA 1570 EUR
APOSOR32122-1G
N-(2-Methylnaphthalen-1-yl)methanesulphonamide, tech
N-(2-Methylnaphthalen-1-yl)methanesulphonamide
Formel: C₁₂H₁₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD03305382
CAS-Nummer: 860610-48-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR