4-(4-Chloro-3-methoxyphenyl)piperidine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321221-100MG
APOSOR321221-100MGEA 434 EUR
APOSOR321221-100MG
4-(4-Chloro-3-methoxyphenyl)piperidine hydrochloride 97%
4-(4-Chloro-3-methoxyphenyl)piperidine hydrochloride
Formel: C₁₂H₁₆ClNO·ClH
Molecular Weight: 262,175 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075580
CAS-Nummer: 1020276-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR