Methyl 2-(piperidin-4-yl)benzoate hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321223-100MG
APOSOR321223-100MGEA 84.1 EUR
APOSOR321223-100MG
Methyl 2-(piperidin-4-yl)benzoate hydrochloride 97%
Methyl 2-(piperidin-4-yl)benzoate hydrochloride
Formel: C₁₃H₁₇NO₂·ClH
Molecular Weight: 255,741 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02179143
CAS-Nummer: 176526-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR