6-(Piperidin-4-yl)-1,2,3,4-tetrahydroquinoline dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321225-100MG
APOSOR321225-100MGEA 168 EUR
APOSOR321225-100MG
6-(Piperidin-4-yl)-1,2,3,4-tetrahydroquinoline dihydrochloride 97%
6-(Piperidin-4-yl)-1,2,3,4-tetrahydroquinoline dihydrochloride
Formel: C₁₄H₂₀N₂·2ClH
Molecular Weight: 289,244 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075584
CAS-Nummer: 2140866-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR