N-(4-(Piperidin-4-yl)pyridin-2-yl)acetamide dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321226-100MG
APOSOR321226-100MGEA 434 EUR
APOSOR321226-100MG
N-(4-(Piperidin-4-yl)pyridin-2-yl)acetamide dihydrochloride 97%
N-(4-(Piperidin-4-yl)pyridin-2-yl)acetamide dihydrochloride
Formel: C₁₂H₁₇N₃O·2ClH
Molecular Weight: 292,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29075585
CAS-Nummer: 1137950-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR