1-(5-(Piperidin-4-yl)indolin-1-yl)ethan-1-one hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR321227-100MG
APOSOR321227-100MGEA 168 EUR
APOSOR321227-100MG
1-(5-(Piperidin-4-yl)indolin-1-yl)ethan-1-one hydrochloride 95%
1-(5-(Piperidin-4-yl)indolin-1-yl)ethan-1-one hydrochloride
Formel: C₁₅H₂₀N₂O·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD29075586
CAS-Nummer: 2244086-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR