(2-(Piperidin-4-yl)phenyl)methanamine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321229-100MG
APOSOR321229-100MGEA 168 EUR
APOSOR321229-100MG
(2-(Piperidin-4-yl)phenyl)methanamine dihydrochloride 97%
(2-(Piperidin-4-yl)phenyl)methanamine dihydrochloride
Formel: C₁₂H₁₈N₂·2ClH
Molecular Weight: 263,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29075589
CAS-Nummer: 2244085-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR