6-(Piperidin-4-yl)isoquinoline dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR321230-100MG
APOSOR321230-100MGEA 79.9 EUR
APOSOR321230-100MG
6-(Piperidin-4-yl)isoquinoline dihydrochloride 95%
6-(Piperidin-4-yl)isoquinoline dihydrochloride
Formel: C₁₄H₁₆N₂·2ClH
Molecular Weight: 285.212 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075590
CAS-Nummer: 2140866-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR