4-(3-Chloro-4-(methylsulphonyl)phenyl)piperidine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321231-100MG
APOSOR321231-100MGEA 346 EUR
APOSOR321231-100MG
4-(3-Chloro-4-(methylsulphonyl)phenyl)piperidine hydrochloride 97%
4-(3-Chloro-4-(methylsulphonyl)phenyl)piperidine hydrochloride
Formel: C₁₂H₁₆ClNO₂S·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD30099236
CAS-Nummer: 1853217-71-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR