2-Methyl-3-(piperidin-4-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321233-100MG
APOSOR321233-100MGEA 125 EUR
APOSOR321233-100MG
2-Methyl-3-(piperidin-4-yl)pyridine dihydrochloride 97%
2-Methyl-3-(piperidin-4-yl)pyridine dihydrochloride
Formel: C₁₁H₁₆N₂·2ClH
Molecular Weight: 249,18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075594
CAS-Nummer: 1256787-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR