(3-(Piperidin-4-yl)phenyl)methanamine dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR321234-100MG
APOSOR321234-100MGEA 168 EUR
APOSOR321234-100MG
(3-(Piperidin-4-yl)phenyl)methanamine dihydrochloride 95%
(3-(Piperidin-4-yl)phenyl)methanamine dihydrochloride
Formel: C₁₂H₁₈N₂·2ClH
Lagertemperatur: Kühlschrank
MDL: MFCD29075596
CAS-Nummer: 864069-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR