2-Methyl-4-(piperidin-4-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321236-100MG
APOSOR321236-100MGEA 346 EUR
APOSOR321236-100MG
2-Methyl-4-(piperidin-4-yl)pyridine dihydrochloride 97%
2-Methyl-4-(piperidin-4-yl)pyridine dihydrochloride
Formel: C₁₁H₁₆N₂·2ClH
Molecular Weight: 249.18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075598
CAS-Nummer: 1020277-65-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR