Ethyl 3-(piperidin-4-yl)picolinate dihydrochloride 90%

Lieferant: Apollo Scientific
Achtung

OR321237-100MG
APOSOR321237-100MGEA 168 EUR
APOSOR321237-100MG
Ethyl 3-(piperidin-4-yl)picolinate dihydrochloride 90%
Ethyl 3-(piperidin-4-yl)picolinate dihydrochloride
Formel: C₁₃H₁₈N₂O₂·2ClH
Molecular Weight: 307.216 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075600
CAS-Nummer: 1853217-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR