2,6-Dimethyl-3-(piperidin-4-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321238-100MG
APOSOR321238-100MGEA 346 EUR
APOSOR321238-100MG
2,6-Dimethyl-3-(piperidin-4-yl)pyridine dihydrochloride 97%
2,6-Dimethyl-3-(piperidin-4-yl)pyridine dihydrochloride
Formel: C₁₂H₁₈N₂·2ClH
Molecular Weight: 263,207 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075601
CAS-Nummer: 2244085-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR