(3-(Piperidin-4-yl)pyridin-2-yl)methanol dihydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR321239-100MG
APOSOR321239-100MGEA
333
EUR
APOSOR321239-100MG
(3-(Piperidin-4-yl)pyridin-2-yl)methanol dihydrochloride 95%
(3-(Piperidin-4-yl)pyridin-2-yl)methanol dihydrochloride
Formel:
C₁₁H₁₆N₂O·2ClH Lagertemperatur: Kühlschrank |
MDL:
MFCD29075602 CAS-Nummer: 1853217-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...