[2-Bromo-5-(hydroxymethyl)-3-methylphenyl]methanol, tech

Lieferant: Apollo Scientific

OR32124-1G
APOSOR32124-1GEA 1570 EUR
APOSOR32124-1G
[2-Bromo-5-(hydroxymethyl)-3-methylphenyl]methanol, tech
[2-Bromo-5-(hydroxymethyl)-3-methylphenyl]methanol
Formel: C₉H₁₁BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03787812
CAS-Nummer: 860610-76-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR