6-(Piperidin-4-yl)-3,4-dihydroisoquinoline-2(1H)-carbaldehyde hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321240-100MG
APOSOR321240-100MGEA 333 EUR
APOSOR321240-100MG
6-(Piperidin-4-yl)-3,4-dihydroisoquinoline-2(1H)-carbaldehyde hydrochloride 97%
6-(Piperidin-4-yl)-3,4-dihydroisoquinoline-2(1H)-carbaldehyde hydrochloride
Formel: C₁₅H₂₀N₂O·ClH
Molecular Weight: 280,793 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075603
CAS-Nummer: 1853217-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR