4-(Piperidin-4-yl)phenol hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR321243-1G OR321243-250MG
APOSOR321243-1GEA 219 EUR
APOSOR321243-1G APOSOR321243-250MG
4-(Piperidin-4-yl)phenol hydrochloride 95%
4-(Piperidin-4-yl)phenol hydrochloride
Formel: C₁₁H₁₅NO·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD10565007
CAS-Nummer: 263139-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR