N-Methyl-1-(4-(piperidin-4-yl)phenyl)methanamine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321244-100MG
APOSOR321244-100MGEA 333 EUR
APOSOR321244-100MG
N-Methyl-1-(4-(piperidin-4-yl)phenyl)methanamine dihydrochloride 97%
N-Methyl-1-(4-(piperidin-4-yl)phenyl)methanamine dihydrochloride
Formel: C₁₃H₂₀N₂·2ClH
Molecular Weight: 277,233 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099243
CAS-Nummer: 1853217-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR