2-Methoxy-4-(piperidin-4-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321247-100MG
APOSOR321247-100MGEA 84.1 EUR
APOSOR321247-100MG
2-Methoxy-4-(piperidin-4-yl)pyridine dihydrochloride 97%
2-Methoxy-4-(piperidin-4-yl)pyridine dihydrochloride
Formel: C₁₁H₁₆N₂O·2ClH
Molecular Weight: 265,179 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099249
CAS-Nummer: 1137950-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR