2-Chloro-1',2',3',6'-tetrahydro-3,4'-bipyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Gefahr

OR321248-100MG
APOSOR321248-100MGEA 168 EUR
APOSOR321248-100MG
2-Chloro-1',2',3',6'-tetrahydro-3,4'-bipyridine dihydrochloride 97%
2-Chloro-1',2',3',6'-tetrahydro-3,4'-bipyridine dihydrochloride
Formel: C₁₀H₁₁ClN₂·2ClH
Molecular Weight: 267,583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30099233
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR