6-(Piperidin-4-yl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride 90%

Lieferant: Apollo Scientific
Achtung

OR321249-100MG
APOSOR321249-100MGEA 333 EUR
APOSOR321249-100MG
6-(Piperidin-4-yl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride 90%
6-(Piperidin-4-yl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
Formel: C₁₄H₂₀N₂·2ClH
Molecular Weight: 289,244 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099234
CAS-Nummer: 1853217-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR