7-(4-Chlorophenyl)-2-methyl-[1,2,4]triazolo[1,5-a]pyridine-8-carbonitrile, tech

Lieferant: Apollo Scientific

OR32125-1G
APOSOR32125-1GEA 1570 EUR
APOSOR32125-1G
7-(4-Chlorophenyl)-2-methyl-[1,2,4]triazolo[1,5-a]pyridine-8-carbonitrile, tech
7-(4-Chlorophenyl)-2-methyl-[1,2,4]triazolo[1,5-a]pyridine-8-carbonitrile
Formel: C₁₄H₉ClN₄
Molecular Weight: 268,701 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787167
CAS-Nummer: 860610-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR