N-Methyl-1-(3-(piperidin-4-yl)phenyl)methanamine dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR321251-100MG
APOSOR321251-100MGEA 333 EUR
APOSOR321251-100MG
N-Methyl-1-(3-(piperidin-4-yl)phenyl)methanamine dihydrochloride 95%
N-Methyl-1-(3-(piperidin-4-yl)phenyl)methanamine dihydrochloride
Formel: C₁₃H₂₀N₂·2ClH
Molecular Weight: 277,233 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099252
CAS-Nummer: 1853217-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR