N,N-Dimethyl-1-(4-(piperidin-4-yl)phenyl)methanamine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321252-100MG
APOSOR321252-100MGEA 333 EUR
APOSOR321252-100MG
N,N-Dimethyl-1-(4-(piperidin-4-yl)phenyl)methanamine dihydrochloride 97%
N,N-Dimethyl-1-(4-(piperidin-4-yl)phenyl)methanamine dihydrochloride
Formel: C₁₄H₂₂N₂·2ClH
Lagertemperatur: Kühlschrank
MDL: MFCD30099253
CAS-Nummer: 2244087-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR