N,N-Dimethyl-1-(2-(piperidin-4-yl)phenyl)methanamine dihydrochloride 90%

Lieferant: Apollo Scientific
Achtung

OR321253-100MG
APOSOR321253-100MGEA 333 EUR
APOSOR321253-100MG
N,N-Dimethyl-1-(2-(piperidin-4-yl)phenyl)methanamine dihydrochloride 90%
N,N-Dimethyl-1-(2-(piperidin-4-yl)phenyl)methanamine dihydrochloride
Formel: C₁₄H₂₂N₂·2ClH
Molecular Weight: 291,26 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099254
CAS-Nummer: 2244087-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR