N-Methyl-1-(2-(piperidin-4-yl)phenyl)methanamine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321254-100MG
APOSOR321254-100MGEA 333 EUR
APOSOR321254-100MG
N-Methyl-1-(2-(piperidin-4-yl)phenyl)methanamine dihydrochloride 97%
N-Methyl-1-(2-(piperidin-4-yl)phenyl)methanamine dihydrochloride
Formel: C₁₃H₂₀N₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD30099255
CAS-Nummer: 1853217-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR