5-(Piperidin-4-yl)nicotinonitrile dihydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321255-100MG
APOSOR321255-100MGEA
433
EUR
APOSOR321255-100MG
5-(Piperidin-4-yl)nicotinonitrile dihydrochloride 97%
5-(Piperidin-4-yl)nicotinonitrile dihydrochloride
Formel:
C₁₁H₁₃N₃·2ClH Molecular Weight: 260,163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30099256 CAS-Nummer: 1137949-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...